missing translation for 'onlineSavingsMsg'
Learn More

SURVIVIN Mouse anti-Human, Clone: 3B9H7, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66495-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16848425

  • 1415.13 NOK / 20µL
Estimated Shipment: 16-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Survivin (IAP4) is the smallest member of the Inhibitors of Apoptosis Protein (IAP) gene family. The IAPs are involved in multiple cell functions such as cell signaling, cell division, metabolism, and exhibits differential expression in nearly all human cancers, but not in most normal tissues. IAP family members usually contain multiple baculovirus IAP repeat (BIR) domains, but the Survivin gene encodes proteins with only a single BIR domain. The encoded proteins also lack a C-terminus RING finger domain. Gene expression is high during fetal development and in most tumors yet low in adult tissues. Antisense transcripts are involved in the regulation of survivin′s gene expression. Survivin is expressed in the G2/M phase of the cell cycle in a cycle-regulated manner. At the beginning of mitosis, survivin associates with microtubules of the mitotic spindle in a specific and saturable reaction that is regulated by microtubule dynamics. Disruption of survivin-microtubule interactions results in loss of survivin′s anti-apoptosis function and increased caspase-3 activity, a mechanism involved in cell death, during mitosis. Survivin may counteract a default induction of apoptosis in G2/M phase. Survivin is also abundantly expressed in brain tissues (astrocytes and some neurons) of adult rats following traumatic brain injury. Survivin has been found co-expressed with NeuN (mature neuronal marker) and PCNA (a cell cycle protein). Survivin might affect regulation of neural cell proliferative responses after brain injury. At least four transcript variants encoding distinct isoforms have been found for this gene, but the full-length natures of only three of them have been determined. The overexpression of survivin in cancer may overcome this apoptotic checkpoint and favor aberrant progression of transformed cells through mitosis.
Specifications

Specifications

SURVIVIN
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O15392
BIRC5
SURVIVIN Fusion Protein Ag20958
20 μL
Primary
Human
Antibody
IgG1
Western Blot
3B9H7
Unconjugated
BIRC5
API4, Apoptosis inhibitor 4, Apoptosis inhibitor survivin, BIRC5, EPR 1, IAP4, SURVIVIN
Mouse
Protein A
RUO
332
-20°C
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers