Learn More
WTW™ Surfactants Test Kits
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
Brand: WTW™ 252062
Additional Details : Weight : 0.80000kg Transport : UN number : 3316 Chem class : 9 Pack group : III
Specifications
For waste water, seepage water, surface water | |
Certificates | |
0.05 to 1.50 mg/L | |
Photometric |
PhotoLab™ S12, PhotoLab™ Spectral photometers | |
25 | |
25 Tests | |
c-Ten (cationic) surfactant cell test kit |