Learn More
Thermo Scientific™ SureSTART™ 9 mm Screw Caps, Level 2 High-throughput Applications
Product Code. p-7236613 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Black
Blue
Green
Red
Yellow
Septum:
Red Silicone/Ivory PFTE
Red Silicone/Ivory PTFE
Red Silicone/White PTFE
White Silicone/Beige PTFE
White Silicone/Red PTFE
White Silicone/Red Polyimide
Product Type:
AVCS Screw Cap
Screw Cap
Quantity:
100 Pk.
1000 Pk.
5000 Pk.
Unit Size:
Pack of 100
Pack of 1000
Pack of 5000
Product Code. Color Septum Product Type Quantity unitSize
17323863 Blue Red Silicone/Ivory PTFE AVCS Screw Cap 100 Pk. Pack of 100
17933611 Blue White Silicone/Red Polyimide AVCS Screw Cap 100 Pk. Pack of 100
17333863 Blue Red Silicone/Ivory PTFE AVCS Screw Cap 5000 Pk. Pack of 5000
17343863 Red Red Silicone/White PTFE AVCS Screw Cap 100 Pk. Pack of 100
17353863 Red Red Silicone/White PTFE AVCS Screw Cap 5000 Pk. Pack of 5000
17363863 Red Red Silicone/White PTFE AVCS Screw Cap 1000 Pk. Pack of 1000
17373863 Blue Red Silicone/Ivory PTFE AVCS Screw Cap 1000 Pk. Pack of 1000
17383863 Blue White Silicone/Red PTFE AVCS Screw Cap 100 Pk. Pack of 100
17393863 Blue White Silicone/Red PTFE AVCS Screw Cap 5000 Pk. Pack of 5000
17303873 Red White Silicone/Red PTFE AVCS Screw Cap 100 Pk. Pack of 100
17313873 Red White Silicone/Red PTFE AVCS Screw Cap 5000 Pk. Pack of 5000
17323873 Red White Silicone/Red PTFE AVCS Screw Cap 1000 Pk. Pack of 1000
17333873 Blue White Silicone/Red PTFE AVCS Screw Cap 1000 Pk. Pack of 1000
17343873 Blue White Silicone/Beige PTFE Screw Cap 100 Pk. Pack of 100
17353873 Blue White Silicone/Beige PTFE Screw Cap 5000 Pk. Pack of 5000
17363873 Blue White Silicone/Beige PTFE Screw Cap 1000 Pk. Pack of 1000
17373873 Blue White Silicone/Beige PTFE Screw Cap 100 Pk. Pack of 100
17383873 Blue White Silicone/Beige PTFE Screw Cap 5000 Pk. Pack of 5000
17393873 Blue White Silicone/Beige PTFE Screw Cap 1000 Pk. Pack of 1000
17805174 Blue Red Silicone/Ivory PFTE AVCS Screw Cap 100 Pk. Pack of 100
17825204 Black White Silicone/Red PTFE AVCS Screw Cap 100 Pk. Pack of 100
17815214 Green White Silicone/Red PTFE AVCS Screw Cap 100 Pk. Pack of 100
17875224 Green Red Silicone/Ivory PTFE AVCS Screw Cap 100 Pk. Pack of 100
17895174 Yellow White Silicone/Red PTFE AVCS Screw Cap 100 Pk. Pack of 100
17875194 Black Red Silicone/Ivory PTFE AVCS Screw Cap 100 Pk. Pack of 100
17895204 Yellow Red Silicone/Ivory PTFE AVCS Screw Cap 100 Pk. Pack of 100
26 options
Product Code. 17323863
Brand: Thermo Scientific™ 6ASC9RT1
Only null left
Add to basket
Edge
Add to basket
Use Level 2 Thermo Scientific™ SureSTART™ 9 mm Screw Caps with screw vials that have a 9 mm opening for your high-throughput applications, when robustness and reproducibility are key.
Use Thermo Scientific™ SureSTART™ 9 mm Screw Caps with screw vials that have a 9 mm opening, including our SureSTART 2 mL Glass Screw Vials and 1.7 mL Glass High Recovery Screw Microvials for <2 mL samples (Level 2). Select Level 2 products for high-throughput applications, when robustness and reproducibility are key. Choose from polypropylene caps with all-purpose silicone/PTFE septa of various hardness values designed to reduce autosampler needle issues. Pre-slit and bonded septa options are available. Our AVCS™ caps incorporate Advanced Vial Closure System technology to ensure optimal seal compression when closing a vial. Use AVCS screw caps with our SureStop™ vials for effective seals, consistent data, and increased sample throughput.
Our performance Level 2 caps are manufactured and tested to ensure low bleeding, robustness, and reproducibility of results for your routine GC-MS and LC-MS analyses.
SureStop Vials & Advanced Vial Closure System (AVCS) Caps:
This patented screw top vial/cap combination features a push-through resistant sealing ring design, controlled septum compression, and SureStop closure positioning flange to ensure an effective seal that eliminates under- and over-sealing, reduces solvent evaporation, and creates less autosampler interruptions. SureStop vials and AVCS caps have universal instrument compatibility for all LC and GC autosamplers. They also contain low leachable levels, making them an ideal alternative to bonded caps for those seeking cleaner spectra.
The SureSTART Performance Levels
SureSTART vials, caps, inserts, septa, and kits are available in three performance levels. Now you can choose the products that best suit your analytical, performance, or budget needs.
• Level 1: Everyday analysis
Chromatography approved
SureSTART Level 1 products are cost optimized, suitable for all instrument types, and quality tested to deliver the performance, sample security, and reproducibility you need for everyday analyses. They are an ideal solution for verifying compound presence and quantification using detection techniques such as UV, RI, and FID.
• Level 2: High-throughput analysis
When robustness and reproducibility are key
SureSTART Level 2 vials have very low metal contents and leachables to ensure that they conform to the high standards you need to meet your regulatory requirements. The septa are conditioned to provide a very low level of chromatographic extractables. These products support routine analyses and deliver high quality data with an uninterrupted workflow.
• Level 3: High performance
When sensitivity is a must
SureSTART Level 3 products are packed in a clean environment according to our own hygiene concept and regulations to protect against contaminating factors. The vials contain the lowest levels of extractable metals and leachables to ensure consistent, reproducible sensitivity and selectivity for the most challenging analytes. The septa are conditioned to provide the lowest level of chromatographic extractables. Choose these products for ultimate method performance and high levels of and sample security and integrity.
All SureSTART vials, closures, and septa are certified to meet physical parameter specifications as part of our manufacturing process.
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Material Polypropylene
Pre-slit No
Septum Red Silicone/Ivory PTFE
Level Level 2
Hardness 55 Shore
Product Line SureSTART Level 2
Quantity 100 Pk.
Thickness (Metric) 1 mm
Closure Size 9 mm
For Use With (Equipment) 9 mm Screw Vials
Certifications/Compliance Specification Certified
Product Type AVCS Screw Cap