Learn More
OHAUS™ Support Stand Universal-H
Shop All Ohaus Products
Click to view available options
Dimensions (L x W x H):
105.000 x 30.000 x 7.000 cm
Unit Size:
1 unit
Description
- Overhead Stirrer Accessory
Specifications
Specifications
| Height (Metric) | 7.000 cm |
| Width (Metric) | 30.000 cm |
| Weight (Metric) | 3.900 kg |
| Dimensions (L x W x H) | 105.000 x 30.000 x 7.000 cm |