Learn More
OHAUS™ Support Stand
Product Code. 15782379 Shop All Ohaus Products
Change view
Click to view available options
Diameter (Metric) Rod:
16 mm
17 mm
18 mm
19 mm
20 mm
21 mm
22 mm
Height (Metric):
267 mm
305 mm
Unit Size:
Each
Product Code. Diameter (Metric) Rod Height (Metric) unitSize
15782379 16 mm 305 mm Each
15722449 16 mm 267 mm Each
15732449 16 mm 267 mm Each
15742449 16 mm 267 mm Each
15752449 16 mm 267 mm Each
15762449 16 mm 267 mm Each
15772449 16 mm 267 mm Each
15792379 17 mm 305 mm Each
15702389 18 mm 305 mm Each
15712389 19 mm 305 mm Each
15722389 20 mm 305 mm Each
15732389 21 mm 305 mm Each
15742389 22 mm 305 mm Each
13 options
Product Code. 15782379
Brand: OHAUS™ 30392334
Only null left
Add to basket
Add to basket
OHAUS™ Support Stand provides secure support for holding glassware at elevated positions.
  • Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
  • Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
  • Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel
Height (Metric) 305 mm
Length (English) 12 in.
Length (Metric) 305 mm
Length (Metric) Rod 457 mm
Width (English) 16.50 in.
Width (Metric) 419 mm
Diameter (Metric) Rod 16 mm
Product Type Support Stand with Rod