Learn More
DWK Life Sciences Kimble™ Ultra-Ware™ PTFE Support Screen Gasket
Click to view available options
Material:
PTFE
Unit Size:
Pack of 25
Description
- Product Type: Support Screen Gasket
- Material: PTFE
- Size: 25 mm
- For Use with: Kimble™ Ultra-Ware™ Microfiltration Assembly
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Disposable | Reusable |
| For Use With (Equipment) | Kimble Ultra-Ware Microfiltration Assembly |
| Material | PTFE |
| No. per Pack | 25 Pk. |
| Type | Gasket |
| Diameter (Metric) | 25 mm |