Learn More
OHAUS™ Support Plate
Product Code. 15732399 Shop All Ohaus Products
Change view
Click to view available options
Dimensions (L x W):
241 x 165 mm
324 x 229 mm
400 x 305 mm
Unit Size:
Each
Product Code. Dimensions (L x W) unitSize
15732399 241 x 165 mm Each
15742399 324 x 229 mm Each
15752399 400 x 305 mm Each
3 options
Product Code. 15732399
Brand: OHAUS™ undefined
Only null left
Add to basket
Add to basket
OHAUS™ Support Plate provides secure support for holding glassware at elevated positions.
  • Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
  • Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
  • Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
TRUSTED_SUSTAINABILITY
Specifications
Material Aluminum
Dimensions (L x W) 241 x 165 mm
Length (Metric) 241 mm
Width (Metric) 165 mm
Product Type Support Plate
Size Small