Learn More
3M™ Supplied Air Respirator System, 06972
Product Code. 31297052 Shop All 3M Products
Change view
Click to view available options
Material:
Polypropylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material
31297052 Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31297052 Supplier 3M™ Supplier No. 7100361625
  • ONLINE EXCLUSIVE PRICE
    13495.00 NOK
    8642.98 NOK / Pack of 1
    Save 4852.02 NOK
    35% Off
Estimated Shipment: 04-12-2026
Provides respiratory protection against particles, gases and vapours, and combination hazards when used with an approved 3M face mask.
  • Compatible with 3M™ Full Facepiece 6000 Series and Half Face Mask 7500 Series.
  • Protects against particles, gases, vapours, and combination hazards.
  • Delivers constant airflow for enhanced comfort.
  • Double-action couplings help prevent accidental disconnection.
  • Adjustable airflow with Flow Check Bag for pre-use testing.
  • Low-profile breathing tube allows free movement and minimal interference.
  • Belt-mounted design with stainless-steel connection for durability and corrosion resistance.
  • ATEX-tested for safe use in IIB 2GD environments.
  • CE approved and compliant with EN14594:2005 standards.
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Product Type Supplied Air Regulator
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Painting, Sanding, Welding
For Use With (Equipment) 3M Face Masks
Model S200+