Learn More
Epredia™ SuperFrost Plus™ Adhesion slides
Product Code. 15438060 Shop All Epredia Products
Change view
Click to view available options
Color:
Blue
Green
Orange
Pink
White
Yellow
Corner Style:
Clipped
Traditional
Unit Size:
Pack of 72
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Corner Style Unit Size
15438060 White Clipped Pack of 72
10149870 White Traditional Pack of 72
12312148 Yellow Traditional Pack of 72
11950657 Blue Traditional Pack of 72
10354365 Pink Traditional Pack of 72
12625336 Green Traditional Pack of 72
15264653 Orange Clipped Pack of 72
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 15438060 Supplier Epredia™ Supplier No. J7800AMNZ
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Minimizes tissue loss during staining resulting in quick turn-around times and reduced risk associated with tissue loss and repeat testing. Epredia™ SuperFrost Plus Slides secure frozen and formalin-fixed, paraffin embedded tissue sections as well as cytology preparations.
  • No time wasted on slide-coating procedures
  • No money wasted on expensive adhesives
  • Reliable and uniform coating
  • Minimal sample loss
  • Reliable and safe adhesive action
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 75 mm
Width (Metric) 25 mm
Edges 90°, Ground
Type Adhesion Slide
Color White
Corner Style Clipped
Quantity 72/Pk.