Learn More
Epredia™ SuperFrost™ Microscope Slides
Product Code. 17294884
Change view
Click to view available options
Color:
Blue
Green
Pink
Yellow
Unit Size:
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17294884 Blue Pack of 50
17204894 Pink Pack of 50
17214894 Yellow Pack of 50
17224894 Green Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17294884 Supplier Epredia™ Supplier No. AG00008132E01MNZ20
Spotlight Offers
Use promo code 28038
View all promotions
 Restricted Item
Estimated Shipment: 14-12-2026
The printed Epredia™ SuperFrost tab is impervious to virtually all common laboratory chemicals and reagents. A wide range of colors allows several examination specimens to be labeled and archived simply and clearly.
  • Time-consuming labeling with paper labels is no longer required
  • The writing area is resistant to all common solvents
  • SuperFrost slides may be sorted by color e.g. by patient, urgency, stain or examination method
  • Simple recognition of specified characteristics such as year of examination or department, use etc.
TRUSTED_SUSTAINABILITY
Specifications
Edges 90°, Ground
Thickness (Metric) 1 mm
Color Blue
Certifications/Compliance ISO8037/I
Coating Uncoated
Writing Surface Yes
Dimensions (L x W) 76 x 26 mm