Learn More
Labcon SuperClear™ Microcentrifuge Tubes
GREENER_CHOICE
Product Code. 16644012
Click to view available options
Color:
Amber
Assorted
Blue
Clear
Green
Natural
Orange
Purple
Red
Yellow
Capacity (Metric):
0.5 mL
1.5 mL
1.7 mL
2.0 mL
Cap Style:
Attached Caps
Extra Large Attached Caps
Snap Cap
Unit Size:
Pack of 1000
Pack of 500
Product Code. 16644012
Brand: Labcon 30343350009
With side graduations, and frosted writing surface
  • Microcentrifuge tube
  • ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
  • Polypropylene USP Class VI resin
  • High centrifuging
  • Freezable to -80°C
  • Made in USA
Specifications
Cap Style Attached Caps
Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Autoclavable Autoclavable
Capacity (Metric) 1.7 mL
Color Natural
Material Polypropylene Resin
For Use With (Application) Centrifugation
Packaging 50 / bag/10 bags / Pk./10 Pk. / case
Clean Claims ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
Sterility Sterile
Product Line SuperClear™ microcentrifuge tubes
Graduated Yes
Max. RCF 15,000 RCF