Learn More
SUN2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27556-1-AP-150UL
This item is not returnable.
View return policy
Description
UNC84B may form a physical interaction between the nuclear envelope and the centrosome.Specifications
| SUN2 | |
| Polyclonal | |
| Unconjugated | |
| SUN2 | |
| FRIGG, KIAA0668, Protein unc 84 homolog B, Rab5 interacting protein, Rab5IP, Sad1/unc 84 protein like 2, SUN2, unc 84 homolog B (C. elegans), UNC84B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 223697, 25777 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8BJS4, Q9UH99 | |
| Sun2 | |
| SUN2 Fusion Protein Ag16077 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |