Learn More
Thermo Scientific™ SUN-SRi™ Assembled 15 x 45mm Screw Thread Vial Kits
Shop All Thermo Scientific ProductsDescription
Pre-assembled kits are convenient for special applications where samples may be injected through the septum. Kits are an economical alternative to ordering multiple components.
Specifications
Specifications
| Closure Color | Black |
| Diameter (English) Outer | 0.5 in. |
| Color | Clear |
| Diameter (Metric) Outer | 15 mm |
| Material | Silica Glass |
| Volume (Metric) | 4 mL |
| Height (English) | 1.7 in. |
| Closure Material | Polypropylene |
| Closure Type | Screw Cap |
| Closure Size | 13-425 |
| Show More |