Learn More
Thermo Scientific™ SUN-SRi™ 13-425 Screw Thread Caps
Product Code. 31920938 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Black
Product Type:
Solid Top Cap
Material:
Phenolic
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Product Type Material Unit Size
31920938 Black Solid Top Cap Phenolic Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31920938 Supplier Thermo Scientific™ Supplier No. 200 568
Estimated Shipment: 21-10-2026
Thermo Scientific caps and vials are available in a wide variety of sizes and configurations to fit the needs of many applications.
TRUSTED_SUSTAINABILITY
Specifications
Material (Liner) PTFE/Rubber
Color Black
Material Phenolic
Closure Size 13-425
Product Type Solid Top Cap