missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech SUMO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10329-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody can detect endogenous levels of SUMOylated proteins (e.g. SUMO-1-RanGAP at 80-90 kDa).
SUMO1 is an ubiquitin-like protein that can be covalently attached to proteins as a monomer or a lysine-linked polymer. Covalent attachment, via an isopeptide bond, to its substrates requires prior activation by the E1 complex SAE1-SAE2 and linkage to the E2 enzyme UBE2I, and can be promoted by E3 ligases such as PIAS1-4, RANBP2 or CBX4. This post-translational modification on lysine residues of proteins plays a crucial role in a number of cellular processes such as nuclear transport, DNA replication and repair, mitosis and signal transduction. SUMO1 is involved, for instance, in targeting RANGAP1 to the nuclear pore complex protein RANBP2. Polymeric SUMO1 chains are also susceptible to polyubiquitination which functions as a signal for proteasomal degradation of modified proteins. SUMO1 may also regulate a network of genes involved in palate development. Mutations in the gene can result in non-syndromic orofacial cleft 10.Specifications
SUMO1 | |
Polyclonal | |
Unconjugated | |
SUMO1 | |
DAP 1, GAP modifying protein 1, GMP1, OFC10, PIC1, SENP2, Sentrin, SMT3, SMT3 homolog 3, SMT3C, SMT3H3, SUMO 1, SUMO1, Ubiquitin like protein SMT3C, Ubiquitin like protein UBL1, UBL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22218, 301442, 7341 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P63165, P63166, Q5I0H3 | |
Sumo1 | |
SUMO1 Fusion Protein Ag0414 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |