missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This antibody detected a major band around 35 kDa in lysates of mouse kidney and human liver.
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene encodes a protein that transfers a sulfo moiety to and from estrone, which may control levels of estrogen receptors.
Specifications
Specifications
| Antigen | SULT1E1 |
| Applications | Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.38 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | SULT1E1 |
| Gene Accession No. | P49888, P49891, P52844 |
| Gene Alias | EST, EST 1, Estrogen sulfotransferase, ST1E1, STE, Sulfotransferase 1E1, SULT1E1 |
| Gene Symbols | Sult1e1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?