missing translation for 'onlineSavingsMsg'
Learn More

Sulphur in Hydrocarbons, Jet Aviation Fuel, 250mL (15.2 mg/kg), Paragon Scientific™

Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for Mercaptan Sulphur Content ISO 3012

Brand:  Paragon Scientific CRM-SUKR

Additional Details : CAS Number : 91770-15-9 Weight : 0.25000kg Transport : UN number : 1863 Chem class : 3 Pack group : III


Product Code. 16070040

  • 2685.00 NOK / 250mL
Estimated Shipment: 11-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Manufactured in accordance with ASTM / EN or IP test methods
  • Certified by Round Robin Method by independant test laboratories
  • Ensures good interlaboratory correlation
  • Supplied in tamper evident UN compliant security pack
  • Pack size 250 mL
  • 12 months shelf life
Specifications

Specifications

1863
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P370+P378-In case of fire: Use media other than water to extinguish.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Nominal values quoted. Certified Values will vary from batch to batch.