Learn More
Sulphate Standard Solution Concentrate, 10ppm, Reagecon™
BIOPHARMA
Product Code. 12564376 Shop All Reagecon Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. 12564376
Brand: Reagecon 5002800C