missing translation for 'onlineSavingsMsg'
Learn More

Sulindac sulfone, ≥99%, MP Biomedicals™

Product Code. 11462661 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462661 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11462661 Supplier MP Biomedicals Supplier No. 0219603005

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 59864-04-9
Color Yellow
Assay Percent Range ≥99%
Molecular Formula C20H17FO4S
Quantity 5 mg
Synonym z-5-fluoro-2-methyl-1-p-methylsulfonyl benzylidene indene-3-acetic acid, sulindac sulfone tlc , solid or powder, 1-4-methylsulfonyl benzylidene-2-methyl-5-fluoro-1h-indene-3-acetic acid, 2-3e-6-fluoro-2-methyl-3-4-methylsulfonylphenyl methylidene inden-1-yl acetic acid
InChI Key MVGSNCBCUWPVDA-RQZCQDPDSA-N
SMILES CC1=C(C2=C(C1=CC3=CC=C(C=C3)S(=O)(=O)C)C=CC(=C2)F)CC(=O)O
IUPAC Name 2-[(3E)-6-fluoro-2-methyl-3-[(4-methylsulfonylphenyl)methylidene]inden-1-yl]acetic acid
Molecular Weight (g/mol) 372.41
PubChem CID 6158261
Formula Weight 372.4
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material Sulindac sulfone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.