Learn More
Sulfuric Acid, Concentrated, 0.05M (0.1N), Scharlab
Product Code. 11352555
Change view
Click to view available options
Quantity:
80 mL
Packaging:
HDPE Ampuole
Unit Size:
1 ampoule
Product Code. Quantity Packaging unitSize
11352555 80 mL HDPE Ampuole 1 ampoule
1 options
Product Code. 11352555
Brand: Scharlab AC207200PA
Eliminate the preparation work. Sulfuric Acid, Concentrated, 0.05M (0.1N), Scharlab is a concentrated volumetric solution with a highly accurate and reliable concentration that can be used to prepare 1L of solution for titration.
  • Packaged in chemically resistant and highly durable plastic ampoules
  • Ampoules contain the precise quantity of concentrate to prepare 1L of ready-to-use standard
  • Obtain different concentrations by diluting the concentrate to a volume other than 1L
  • Concentrated solution saves on shipping costs and storage space
Specifications
Color Colorless
CAS 7664-93-9
Composition 4.904g H2SO4
Boiling Point 135°C
Concentration 0.5mol/L in concentrate (0.05mol/L prepared)
Density 1.06g/cm3
Packaging HDPE Ampuole
Normality 0.1N (prepared)
Quantity 80 mL
Molecular Formula H2O4S
Physical Form Liquid
MDL Number MFCD00064589
UN Number 2796
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
Solubility Information Miscible in water
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
SMILES OS(O)(=O)=O
IUPAC Name sulfuric acid
Molecular Weight (g/mol) 98.07
PubChem CID 1118
ChEBI CHEBI:26836
Formula Weight 98.08g/mol
Chemical Name or Material Sulfuric acid
Grade Applications: analytical chemistry, laboratory reagent, titrant in volumetric analysis