missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 95-97%, Reagent Grade, ISO, Ph Eur, Scharlab

Product Code. 11392545
Change view
Click to view available options
Quantity:
25 L
Packaging:
HDPE Carboy
Unit Size:
25 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11392545 25 L HDPE Carboy 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11392545 Supplier Scharlab Supplier No. AC2067025P
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Complies with International Standards Organization and European Pharmacopeia specifications. Sulfuric Acid, 95-97%, Reagent Grade, ISO, Ph Eur, Scharlab is packed in HDPE carboy for great resistance and durability to ensure product safety and quality.

Reagent grade solutions are the most frequently used grade for laboratory analytical work. They correspond to a grade of high quality chemicals for laboratory and specialized industrial use.

Specifications

Appearance Clear, colorless
CAS 7664-93-9
Melting Point -15°C
Boiling Point 310°C
Density 1.834 - 1.837g/cm3
Heavy Metals (as Pb) Max. 0.0001%
Packaging HDPE Carboy
Residue after Ignition Max. 0.0003%
Vapor Pressure 0.0001 hPa
Assay Percent Range 95-97%
Quantity 25 L
Molecular Formula H2O4S
Physical Form Liquid
MDL Number MFCD00064589
UN Number 1830
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
Solubility Information Miscible in water
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
SMILES OS(O)(=O)=O
IUPAC Name sulfuric acid
Molecular Weight (g/mol) 98.07
PubChem CID 1118
ChEBI CHEBI:26836
Formula Weight 98.08g/mol
Grade ISO, Reag. Ph Eur
Chemical Name or Material Sulfuric acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.