Learn More
Sulfuric Acid, 2.5M/l Solution, Scharlab
Product Code. 11392555
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11392555
Brand: Scharlab AC20861000
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Used in analytical chemistry, as a laboratory reagent and a neutralizing agent.
This volumetric solution was checked by means of potentiometric methods using Scharlau's tris(hydroxymethyl)- aminomethane volumetric standard. Scharlau's volumetric standards are directly traceable to the Standard Reference Materials from NIST (National Institute of Standards and Technology, USA).
  • Synonym: Sulphuric acid
  • Molecular Formula: H2SO4
  • Molar Mass: 98.08g/mol
  • CAS: 7664-93-9
  • EIN ECS No.: 231-639-5
  • Factor: 0.999–1.001
Specifications
CAS 7664-93-9
Boiling Point ∼103°C
Concentration 2.5mol/L
Density 1.15g/cm3 
Traceability to NIST Traceable to NIST
Normality 5N
Quantity 1 L
Molecular Formula H2O4S
Physical Form Solution
MDL Number MFCD00064589
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
SMILES OS(O)(=O)=O
IUPAC Name sulfuric acid
Molecular Weight (g/mol) 98.07
PubChem CID 1118
ChEBI CHEBI:26836
Formula Weight 98.08g/mol
Show More Show Less
compliance-icons
Product Identifier
  • Sulfuric acid, solution 2,5 mol/L (5N)
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Sulphuric acid