missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 10% w/v, Extra Pure, Scharlab
Description
- Synonym: Sulphuric acid
- Molecular Formula: H2SO4
- Molar Mass: 98.08g/mol
- CAS: 7664-93-9
- EIN ECS No.: 231-639-5
Specifications
Specifications
| CAS | 7664-93-9 |
| Density | 1.06g/cm3 |
| Residue after Ignition | 0.0005% max |
| Quantity | 1 L |
| Molecular Formula | H2O4S |
| Physical Form | Solution |
| MDL Number | MFCD00064589 |
| Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
| InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
| SMILES | OS(O)(=O)=O |
| Show More |
Safety and Handling
Product Identifier
- Sulfuric acid, solution 10% w/v, extra pure
- Danger
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P362-Take off contaminated clothing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Sulphuric Acid
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?