Learn More
Sulfuric Acid, 0.02M, Chem-Lab
Product Code. 10602913
Change view
Click to view available options
Quantity:
1 L
Packaging:
Polyethylene
Unit Size:
1 L
Product Code. Quantity Packaging unitSize
10602913 1 L Polyethylene 1 L
1 options
Product Code. 10602913
Brand: Chem-Lab CL05.2652.1000
Ideal for titration and TMAR TA-TAC determination. Sulfuric Acid, 0.02M, Chem-Lab is a solution of 1.962g H2SO4 per liter of water for use in laboratory applications.
Sulfuric acid is a highly corrosive, strong mineral acid that shows different properties depending upon its concentration. It is a pungent-ethereal, colorless to slightly yellow viscous liquid. Sulfuric acid is a diprotic acid that is hygroscopic. Concentrated sulfuric acid has strong dehydrating and oxidizing properties. Dilute sulfuric acid is a good electrolyte.
Specifications
pH <2
Color Colorless
CAS 7664-93-9, 7732-18-5
Composition 1.962g H2SO4 per 1L H2O
Melting Point 0°C
Boiling Point 100°C
Concentration 0.02mol/L
Density 1.00g/mL
Odor Odorless
Packaging Polyethylene
Normality 0.04N (±0.00008, 20°C)
Quantity 1 L
Molecular Formula H2O4S
Viscosity 1 cSt
Physical Form Liquid
MDL Number MFCD00064589
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
Solubility Information Soluble in Water
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
SMILES OS(O)(=O)=O
IUPAC Name sulfuric acid
Molecular Weight (g/mol) 98.07
PubChem CID 1118
ChEBI CHEBI:26836
Formula Weight 98.08g/mol
Show More Show Less
compliance-icons
Product Identifier
  • Sulfuric acid 0.02 mol/l
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.