Learn More
Sulfolane, Sigma-Aldrich™
Product Code. 15899530
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15899530 1 L 1 L
15826470 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15899530 Supplier Sigma Aldrich Supplier No. 8079931000
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 126-33-0
Specifications
Melting Point 26°C
Density 1.26 g/cm3 at 30°C
Boiling Point 285°C/1013 hPa
MDL Number MFCD00005484
Quantity 1 L
Synonym Sulfolane, Tetrahydrothiophene 1,1-dioxide, Tetramethylene sulfone, Sulfolane
Molecular Weight (g/mol) 120.17
Grade Research