missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfo-SMCC sodium, MedChemExpress
Description
Sulfo-SMCC sodium is a commonly used hetero-bifunctional, noncleavable ADC crosslinker bearing N-hydroxysuccinimide (NHS) ester and maleimide groups to react with primary amines and sulfhydryl groups, respectively.
Specifications
Specifications
| Chemical Name or Material | Sulfo-SMCC sodium |
| CAS | 92921-24-9 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C16H17N2NaO9S |
| Quantity | 100 mg |
| Solubility Information | DMSO : 125 mg/mL (286.45 mM; Need ultrasonic) ∣H2O : 5 mg/mL (11.46 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | O=C(C1CCC(CN2C(C=CC2=O)=O)CC1)ON3C(C(S(=O)(O[Na])=O)CC3=O)=O |
| Molecular Weight (g/mol) | 436.37 |
| Formula Weight | 436.37 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?