Learn More
WTW™ Sulfite Reagent Test Kit
Product Code. 11727934
Click to view available options
No. of Tests:
150
Unit Size:
Each
Product Code. 11727934
Brand: WTW™ 252053
Only null left
Add to basket
Add to basket
With certificate for optimum precision.
  • Convenient and cost-effective
  • Assured quality by AQA/IQC
  • For low concentrations and routine work with large sample throughput in the laboratory
  • Wide measuring range using rectangular cuvettes for determination of trace concentrations
TRUSTED_SUSTAINABILITY
Specifications
Analytes Chromosulfuric Acid oxidation
For Use With (Application) For waste water, boiler water, foodstuffs
For Use With (Equipment) For PhotoLab™ S12, PhotoLab™ Spectral photometers
Includes Certificates
No. of Tests 150
Range 1.0 - 60.0mg/I SO₃
Test Method Photometric