Learn More
Sulfite, Quant™ Test Strips
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Description | 100 strips to measure 10-500ppm SO3⁻2 in aqueous solution. |
| Includes | 1 kit |
| No. per Pack | 100 strips |
| Quantity | 1 Kit |
| Storage Requirements | Ambient temperatures |
| Type | Sulfite, Quant™ Test Strips |
Frequently Asked Questions (FAQs)
The Sulfite, Quant Test Strips can measure sulfite ion in the range of 10 to 500 ppm.
You will receive 100 test strips in each pack of Sulfite, Quant Test Strips.
The Sulfite, Quant Test Strip is a semi-quantitative test for measuring sulfite (SO32-) ion in aqueous solution.