Learn More
Sulfaquinoxaline sodium 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30318044
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
30318044 1 mL 1mL
1 options
Product Code. 30318044
Brand: Dr. Ehrenstorfer DREA16990020AL100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sulfaquinoxaline sodium salt
CAS 967-80-6
Analyte or Component Names Sulfaquinoxaline sodium salt
Shipping Condition -18°C
Molecular Formula C14 H11 N4 O2 S . Na
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Sulfaquinoxaline Sodium
InChI Formula InChI=1S/C14H11N4O2S.Na/c15-10-5-7-11(8-6-10)21(19,20)18-14-9-16-12-3-1-2-4-13(12)17-14;/h1-9H,15H2;/q-1;+1
SMILES [Na+].Nc1ccc(cc1)S(=O)(=O)[N-]c2cnc3ccccc3n2
IUPAC Name sodium;(4-aminophenyl)sulfonyl-quinoxalin-2-ylazanide
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 322.32
Quantity 1 mL
Formula Weight 322.05 g/mol