Learn More
Sulfamonomethoxine-d4, TRC
Product Code. 30418557
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30418557 25 mg -
30419367 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30418557 Supplier Toronto Research Chemicals Supplier No. TRCS69914725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1286538-12-2
Molecular Formula C11H8D4N4O3S
Molecular Weight (g/mol) 284.33
Synonym 4-Amino-N-(6-methoxy-pyrimidin-4-yl)benzenesulfonamide-d4, 4-Methoxy-6-
IUPAC Name 4-amino-N-(6-methoxypyrimidin-4-yl)benzene-2,3,5,6-d4-sulfonamide
SMILES NC(C([2H])=C1[2H])=C([2H])C([2H])=C1S(NC2=CC(OC)=NC=N2)(=O)=O
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C11H12N4O3S/c1-18-11-6-10(13-7-14-11)15-19(16,17)9-4-2-8(12)3-5-9/h2-7H,12H2,1H3,(H,13,14,15)/i2D,3D,4D,5D
Formula Weight 284.09
Percent Purity >95%
Chemical Name or Material Sulfamonomethoxine-d4
Grade HPLC