Learn More
Sulfamic acid, ≥99.5% (alkalimetric), Solstice
Product Code. 15680860 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
25 kg
4 x 5 kg
6 x 2.5 kg
Unit Size:
1kg
2.5kg
25kg
5kg
Pack of 4
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15680860 25 kg 25kg
15660860 1 kg 1kg
15670860 2.5 kg 2.5kg
15600870 5 kg 5kg
15610870 6 x 2.5 kg Pack of 6
15690860 4 x 5 kg Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15680860 Supplier Fluka Supplier No. 0740125KG
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
≥99.5% (alkalimetric)
  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5329-14-6
Molecular Formula H3NO3S
Molecular Weight (g/mol) 97.088
MDL Number MFCD00011603
InChI Key IIACRCGMVDHOTQ-UHFFFAOYSA-N
Synonym amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
PubChem CID 5987
ChEBI CHEBI:9330
IUPAC Name sulfamic acid
SMILES NS(=O)(=O)O
Specifications
CAS Max % ≤100.0000%
Packaging Paper / Poly Bag
Residue after Ignition ≤0.02%
Quantity 25 kg
Linear Formula NH2SO3H
UN Number UN2967
Formula Weight 97.09
Percent Purity >99.5%
Assay alkalimetric
Chemical Name or Material Sulfamic acid, ≥99.5% (alkalimetric)