Learn More
Sulfachloropyridazine-(phenyl-13C6), Honeywell Fluka™
Product Code. 15987402 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
15987402 10 mg 10mg
1 options
Product Code. 15987402
Brand: Honeywell Fluka R101810MG
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Analytical standard, reference material
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY
Specifications
Packaging Glass Bottle
Physical Form Solid
Molecular Formula C413C6H9ClN4O2S
Industry Type Sulfonamides
Molecular Weight (g/mol) 290.68
Quantity 10 mg
Grade Analytical Standard