missing translation for 'onlineSavingsMsg'
Learn More

Sulfachloropyrazine, TRC

Product Code. 17968504
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17968504 100 mg 100mg
30012255 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17968504 Supplier Toronto Research Chemicals Supplier No. TRCS689063100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-65-8
Molecular Formula C10 H9 Cl N4 O2 S
Molecular Weight (g/mol) 284.72
Synonym Benzenesulfonamide, 4-amino-N-(6-chloropyrazinyl)- (9CI), Sulfanilamide, N1-(6-chloropyrazinyl)- (7CI,8CI), 4-Amino-N-(6-chloro-2-pyrazinyl)benzenesulfonamide, Esb3, Esb3 (antibiotic), N1-(6-Chloropyrazinyl)sulfanilamide, Sulfachloropyrazine, Sulfaclozine, Sulfatyf
IUPAC Name 4-amino-N-(6-chloropyrazin-2-yl)benzenesulfonamide
SMILES Nc1ccc(cc1)S(=O)(=O)Nc2cncc(Cl)n2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H9ClN4O2S/c11-9-5-13-6-10(14-9)15-18(16,17)8-3-1-7(12)2-4-8/h1-6H,12H2,(H,14,15)
Formula Weight 284.0135
Percent Purity >95%
Chemical Name or Material Sulfachloropyrazine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.