missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SUG1 Polyclonal antibody specifically detects SUG1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | SUG1 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 610 SE |
| Formulation | 50mM Sodium Borate |
| Gene Alias | MSUG1 protein, p45, p45/SUG26S protease regulatory subunit 8, proteasome (prosome, macropain) 26S subunit, ATPase, 5, proteasome 26S ATPase subunit 5, Proteasome 26S subunit ATPase 5, Proteasome subunit p45, S8, SUG-1, SUG126S proteasome AAA-ATPase subunit RPT6, Tat-binding protein homolog 10, TBP10, thyroid receptor interactor 1, TRIP1Thyroid hormone receptor-interacting protein 1 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 127-406 of human SUG1 (NP_002796.4).,, Sequence:, LPNKVDPLVSLMMVEKVPDSTYEMIGGLDKQIKEIKEVIELPVKHPELFEALGIAQPKGVLLYGPPGTGKTLLARAVAHHTDCTFIRVSGSELVQKFIGEGARMVRELFVMAREHAPSIIFMDEIDSIGSSRLEGGSGGDSEVQRTMLELLNQLDGFEATKNIKVIMATNRIDILDSALLRPGRIDRKIEFPPPNEEARLDILKIHSRKMNLTRGINLRKIAELMPGASGAEVKGVCTEAGMYALRERRVHVTQEDFEMAVAKVMQKDSEKNMSIKKLWK |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?