Learn More
SUDS3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25845-1-AP-20UL
 View more versions of this product
Product Code. 16844405
  • 1710.33 NOK / 20µL
Description
SDS3 is a subunit of the histone deacetylase.
Specifications
Specifications
SUDS3
Polyclonal
Unconjugated
SUDS3
SAP45, SDS3, SUDS3
Rabbit
Antigen Affinity Chromatography
RUO
64426
-20°C
Liquid
Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9H7L9
SUDS3
SUDS3 Fusion Protein Ag22961