Learn More
Hirschmann™ Suction Tube with Recirculation
Click to view available options
Length (Metric):
310 mm
400 mm
2000 mm
5000 mm
Includes:
w/ Recirculation
w/o Recirculation
w/out Recirculation
Unit Size:
Each
Description
- Screw-coupled
- Safely attached to the instrument
Specifications
Specifications
| Product Type | Suction Hose |
| Material | FEP |
| For Use With (Equipment) | Ceramus Classic |
| Includes | w/ Recirculation |
| Length (Metric) | 400 mm |