Learn More
Buerkle™ Suction Heads for Mini ViscoSampler™
Product Code. 10772521 Shop All Bürkle Products
Change view
Click to view available options
Size:
2 mm Drill
4 mm Drill
6 mm Drill
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
10772521 2 mm Drill Each
10544041 4 mm Drill Each
10329920 6 mm Drill Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10772521 Supplier Buerkle™ Supplier No. 53411002
  • ONLINE EXCLUSIVE PRICE
    577.00 NOK
    570.31 NOK / Pack of 1
    Save 6.69 NOK
    1% Off
Estimated Shipment: 29-10-2026
Add to basket
Add to basket
Buerkle™ V4A Stainless Steel Suction Head is a replacable suction head for use with Buerkle™ Mini ViscoSampler.
  • Replaceable suction heads for Mini ViscoSampler
TRUSTED_SUSTAINABILITY
Specifications
Product Type Suction Head
Material Stainless Steel (V4A)
Autoclavable Autoclavable
Description 2 mm Suction Drill
Size 2 mm Drill
For Use With (Equipment) Mini ViscoSampler