Learn More
Sucrose (Brix) Standard, 35%, Reagecon™
Product Code. 13444036 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
Product Code. Quantity unitSize
13444036 15 mL 15mL
1 options
Product Code. 13444036
Brand: Reagecon BS35S