Learn More
Sucrose (Brix) Standard, 25%, Reagecon™
BIOPHARMA
Product Code. 13464026 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
Product Code. Quantity unitSize
13464026 15 mL 15mL
1 options
Product Code. 13464026
Brand: Reagecon BS25S