Learn More
Sucrose Brix/RI Standard, UKAS ISO 17025 / ISO 17034 Certified, Sucrose (11.20 °Brix / 1.34968 RI) Standard, 15 mL, Paragon Scientific™
Product Code. 16010410
Change view
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
Product Code. Quantity unitSize
16010410 15 mL 15mL
1 options
Product Code. 16010410
Brand: Paragon Scientific SS112
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Paragon ISO 17025 and ISO 17034 certified brix standards are for the calibration and verification of all types of refractometer e.g handheld, Abbe and high accuracy digital instruments. Typically used within the food and beverage and pharmaceutical industries.
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Primary level refractive index standards traceable to National Standards
  • Manufactured in accordance with ICUMSA Methods
  • Supplied in tamper evident packs with 6 weeks shelf life
Specifications
Quantity 15 mL
Nominal values quoted. Certified Values will vary from batch to batch.