missing translation for 'onlineSavingsMsg'
Learn More

Succinyl sulfathiazole, 99.48%, MP Biomedicals™

Product Code. p-4774217 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11413351

Brand: MP Biomedicals 0210297680

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Succinylsulfathiazole is used to block the synthesis of folic acid. It is used to study folate deficiency

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-43-8
Molecular Formula C13H13N3O5S2
Molecular Weight (g/mol) 355.383
MDL Number MFCD00022437
InChI Key SKVLYVHULOWXTD-UHFFFAOYSA-N
Synonym succinylsulfathiazole, cremosuxidine, sulfasuccidine, sulfasuccinil, sulfasuxidine, colistatin, sulfadigesin, sulfasuccithiazole, sulfasuccidin, sulfenterone
PubChem CID 5315
ChEBI CHEBI:9309
IUPAC Name 4-oxo-4-[4-(1,3-thiazol-2-ylsulfamoyl)anilino]butanoic acid
SMILES C1=CC(=CC=C1NC(=O)CCC(=O)O)S(=O)(=O)NC2=NC=CS2

Specifications

Color White
Quantity 100 g
Formula Weight 355.4
Physical Form Powder
Chemical Name or Material Succinyl sulfathiazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.