missing translation for 'onlineSavingsMsg'
Learn More

Succinic acid, Free acid, ≥99%, Ultrapure

Product Code. 15855338 Shop All Thermo Scientific Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 15855338

Brand: Thermo Scientific J21900.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 110-15-6 | C4H6O4 | 118.088 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-15-6
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.088
MDL Number MFCD0000278
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
Synonym succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
PubChem CID 1110
ChEBI CHEBI:15741
IUPAC Name butanedioic acid
SMILES C(CC(=O)O)C(=O)O

Specifications

Melting Point 186°C to 188°C
Boiling Point 235°C
Linear Formula HOOCCH2CH2COOH
Beilstein 1754069
Formula Weight 118.09
Percent Purity ≥99%
Density 1.56
Quantity 1 kg
Flash Point >110°C
Packaging Plastic bottle
Physical Form Crystalline Powder
Specific Gravity 1.56
Chemical Name or Material Succinic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.