missing translation for 'onlineSavingsMsg'
Learn More

Succinate, 0.2M buffer soln., pH 5.5, Thermo Scientific™

Product Code. 15403759
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
This item is not returnable. View return policy

Product Code. 15403759

Brand: Thermo Scientific Alfa Aesar J61100.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-90-3
Molecular Formula C4H4Na2O4
Molecular Weight (g/mol) 162.052
MDL Number MFCD00002789
InChI Key ZDQYSKICYIVCPN-UHFFFAOYSA-L
PubChem CID 9020
ChEBI CHEBI:63675
IUPAC Name disodium;butanedioate
SMILES C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+]

Specifications

Color Colorless
Concentration 0.2 M
pH 5.5
Physical Form Liquid
Quantity 500 mL
Odor Odorless
Chemical Name or Material Succinate buffer soln.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.