Learn More
Hoefer™ SUBHT High Throughput Submarine Gel Unit
Ideal for high throughput screening applications like single strand polymorphism (SSP) analysis following PCR
Brand: Hoefer™ SUB-SG
Additional Details : Weight : 0.01000kg
Includes:
- Buffer chamber assembly
- Safety lid, high-voltage leads
- Fluorescent Labeled Running Tray with Casting Gates
- Four 1mm thick 30-well combs
- Colored Loading Strips
- Two Buffer Recirculation Ports
Description
- Fast, error-free loading: load an entire 96-well plate using a multichannel pipetter
- Simplified Analysis: Fluorescent labels for each sample printed on a UV transparent gel tray allows for instant sample identification (matched to a 96-well plate) during post-run visualization
- High Throughput: Run up to 120 samples (96-well plate plus 24 control and/or marker samples) in 30 minutes
- Better Reproducibility: Eliminate variability caused by running buffer pH drift and ionic gradient formation with optional buffer recirculation capability
- Reduced Buffer Needs: Compact tank design needs less buffer to cover gel which not only reduces buffer consumption but provides greater control of voltage gradients and run times
- Maximum Flexibility: Fully compatible with a wide range of combs and accessories to meet all your high throughput and standard gel analysis needs
- This high throughput horizontal gel unit can accommodate up to 120 samples and resolve DNA strands from 50-1000 base pairs in 30 minutes
- Gel Dimensions: 15 x 15 cm
- Maximum Buffer Volume: 1.2L
- Maximum Power Settings: 300V, 200mA, 60W
- Maximum Sample Capacity: 120
- Maximum Running Temperature: 45°C
- Indoor Use: 4-40°C
- Humidity: up to 80%
- Unit Dimensions: 33.5L x 21.5W x 7 cmH
- Safety Certifications: EN61010-1, CE
Applications:
Single Strand Polymorphism (SSP) analysis
Specifications
SUBHT High Throughput Submarine Gel Unit | |
Silicone Gasket | |
Silicon |