Learn More
Memmert Subframe height 130mm (only with corresponding stacking set) for HCP50
Brand:  Memmert HCP50B33507
Subframe height 130mm (only with corresponding stacking set) for HCP50
Product Code. 16364867
  • ONLINE EXCLUSIVE PRICE
    3915.00 NOK
    3556.43 NOK / Each
    Save 358.57 NOK
    9% Off
Only null left
Estimated Shipment: 11-03-2026