Learn More
Styrene, Sigma-Aldrich™
Product Code. 15899520
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Unit Size:
1 L
100mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15899520 100 mL 100mL
15886460 1 L 1 L
15896460 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15899520 Supplier Sigma Aldrich Supplier No. 8076790100
 Restricted Item
Estimated Shipment: 18-11-2026
Stabilized for synthesis
(stabilized) for synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 100-42-5
Specifications
Melting Point -31°C
Density 0.906 g/cm3 at 20°C
Boiling Point 145°C/1013 hPa
MDL Number MFCD00008612
Quantity 100 mL
Synonym Styrene, Phenylethylene, Vinylbenzene, Styrene
Molecular Weight (g/mol) 104.15
Grade Research