Learn More
STXBP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15312-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the STXBP/unc-18/SEC1 family. The encoded protein is involved in intracellular trafficking, control of SNARE (soluble NSF attachment protein receptor) complex assembly, and the release of cytotoxic granules by natural killer cells. Mutations in this gene are associated with familial hemophagocytic lymphohistiocytosis. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.Specifications
| STXBP2 | |
| Polyclonal | |
| Unconjugated | |
| STXBP2 | |
| Hunc18b, MUNC18 2, pp10122, STXBP2, STXBP2 protein, syntaxin binding protein 2, UNC18 2, UNC18B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20911, 6813 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15833, Q64324 | |
| STXBP2 | |
| STXBP2 Fusion Protein Ag7537 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |