Learn More
Cole-Parmer™ Accessories for Cole-Parmer™ SH-200 Series Shakers and SI-200 Series Shaking Incubators
Product Code. p-6837045
Click to view available options
Product Type:
Platform Mat
Tier System
Description:
for Large Orbital Shakers
for Large Rocking Shakers
for Small Rocking Shakers
Unit Size:
Each
Product Code. 10195704
Brand: Cole-Parmer™ 5170094
Only null left
Add to basket
Add to basket
Optimize the performance of your Cole-Parmer™ shaking incubators.
  • Keep your shaking incubator running properly with these accessories.
Specifications
Product Type Tier System
Material Metal
Length (English) 16.5 in.
Length (English) Platform 14 in.
Length (Metric) 420 mm
Length (Metric) Platform 355 mm
Width (English) 14.2 in.
Width (English) Platform 14 in.
Width (Metric) 360 mm
Width (Metric) Platform 355 mm
Description for Large Rocking Shakers
For Use With (Equipment) 3D Gyratory Rocker SSL3, See-Saw Rocker SSL4
Includes 2 platforms trays, 8 bars