Learn More
Cole-Parmer™ Accessories for Cole-Parmer™ SH-200 Series Shakers and SI-200 Series Shaking Incubators
Product Code. 10195704
Change view
Click to view available options
Product Type:
Platform Mat
Tier System
Description:
for Large Orbital Shakers
for Large Rocking Shakers
for Small Rocking Shakers
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Description unitSize
10195704 Tier System for Large Rocking Shakers Each
10227674 Tier System for Small Rocking Shakers Each
12974025 Platform Mat for Large Orbital Shakers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10195704 Supplier Cole-Parmer™ Supplier No. 5170094
Only null left
Add to basket
Add to basket
Optimize the performance of your Cole-Parmer™ shaking incubators.
  • Keep your shaking incubator running properly with these accessories.
Specifications
Product Type Tier System
Material Metal
Length (English) 16.5 in.
Length (English) Platform 14 in.
Length (Metric) 420 mm
Length (Metric) Platform 355 mm
Width (English) 14.2 in.
Width (English) Platform 14 in.
Width (Metric) 360 mm
Width (Metric) Platform 355 mm
Description for Large Rocking Shakers
For Use With (Equipment) 3D Gyratory Rocker SSL3, See-Saw Rocker SSL4
Includes 2 platforms trays, 8 bars
Vis mere Vis mindre
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.