Learn More
Techne™ Stuart™ Stainless Steel Plunger
Product Code. p-8012918
Change view
Click to view available options
Length (Metric):
16 mm
19 mm
22 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
11553829 16 mm Each
11563829 22 mm Each
11543829 19 mm Each
3 options
Product Code. 11553829
Brand: Techne™ 9893049
Only null left
Add to basket
Add to basket
Parts and accessories to keep your gel timer running smooth and accurate.
Specifications
For Use With (Equipment) Gel Timers
For Use With (Application) For Easy Cleaning
Length (Metric) 16 mm
Type Plunger
Material Stainless Steel